Learn More
Aalborg™ Gas Mass Flow Controller
Product Code. 11747879
Click to view available options
Flow Rate Range:
0 to 1 L/min.
0 to 10 mL/min.
0 to 20 L/min.
0 to 30 L/min.
Unit Size:
Each
Product Code. 11747879
Brand: Aalborg™ GFC17AVAL6C0/3AB07SS
Only null left
Add to basket
Add to basket
Designed to indicate and control set flow rates of gases. Aalborg™ Gas Mass Flow Controller combines the characteristics and accuracy of conventional mass flow devices into a unique compact design at low costs previously unattainable.
  • Rigid metallic construction
  • Built-in tiltable LCD readout
  • Circuit protection
  • Can be used as a portable device
  • Engineering units or 0 to 100% displays
  • TIO - Totalizer option
Certifications
NIST
Specifications
Type Flow Controller
Accuracy ±1%
Airflow Rate 0 to 1 L/min.
Gas Type Air
Pressure Rating 25 psig
Connections 0.63 cm (0.25 in.) compression fittings
Description Transports air
Flow Rate Range 0 to 1 L/min.
For Use With (Application) Indicates and controls set flow of gases
Power Supply Details 12 - 26V, 200mA (maximum)
Repeatability ±0.25% (Full Scale)
Temperature (English) 32° to 122°F
Temperature (Metric) 0°C to 50°C