Learn More
Greiner Bio-One Masterblock™ Polypropylene 96-Well Solid U-Bottom Microplates
Brand: Greiner Bio-One 780265
This item is not returnable.
View return policy
Description
Specifications
Standard | |
Green | |
96 | |
Round Bottom (U-well) | |
Microplates for storing sample material but are also ideal for cultivating bacteria or yeasts |
8 x 12 | |
Polypropylene | |
1 mL | |
Yes | |
-196°C to +121°C |