Learn More
Eppendorf™ Mastercycler™ Nexus Flat
Brand: Eppendorf™ 6335000011
This item is not returnable.
View return policy
Description
Specifications
250mm | |
320mm | |
Mastercycler Nexus Flat | |
Aluminum | |
30 to 99°C | |
20°C |
410mm | |
230V | |
50/60Hz | |
3°C/s | |
230V, 50/60Hz, 700w |