Learn More
Fisherbrand™ Floor Mats
Helps prevent fatigue when standing for long durations.
3.10€ - 4.95€
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
12806445
|
Fisherbrand™
2047/2 |
100 x 100 mm |
3.10€
Each |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
12816445
|
Fisherbrand™
2047/5 |
170 x 160 mm |
4.95€
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
- Polymeric compound
- Available in different sizes
- Blue in color
Specifications
1 Ea. | |
Mat | |
Blue |
Product Title
Fisherbrand™ Floor Mats