Learn More
Tocris Bioscience™ MaxiPost
Potassium channel modulator; exerts subtype-specific effects
Brand:  Tocris Bioscience™ 4949/50
 View more versions of this product
Product Code. 18718599
  • 1165.00€ / 50mg
Estimated Shipment: 29-01-2025
Description
MaxiPost is a potassium channel modulator. Acts as a positive modulator at neuronal Kv7 channels and calcium-activated K+ channels (BKCa) in HEK293 cells. Displays negative modulatory activity at Kv7.1 channels (Ki = 3.7 μM) and GABAA receptors. Displays anxiolytic activity in vivo.
Specifications
Specifications
>98%
MaxiPost
MaxiPost
50 mg
359.7