Learn More
Invitrogen™ MDC Human ProQuantum Immunoassay Kit
GREENER_CHOICE
Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument
Brand:  Invitrogen™ A42895
 View more versions of this product
Product Code. 16351994
  • 748.00€ / Pack of 96
Estimated Shipment: 22-01-2025
Description
  • High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
  • Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
  • Fast, easy workflow: No wash steps, 2 hours from sample to answer
  • Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
  • No proprietary instrument to purchase: Runs on any real-time PCR instrument
  • ProQuantum software available: Robust data analysis and statistical group wise comparison
TRUSTED_SUSTAINABILITY
Specifications
Specifications
O00626
<1.5 pg/mL
Proximity-based Immunoassay
Human
qPCR Instruments
1 hr.
FAM™
RUO
Frozen at -20°C
30 min.
1.6 to 25,000 pg/mL
Taqman™ Real-time PCR
ProQuantum
Protein quantitation
6367
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard
96 Tests
Serum,2-5μL; Supernatant,2-5μL; Plasma,2-5μL
Human
2 hr.