Learn More
WTW™ Measuring Bottle
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
Brand:  WTW™ 209106
Product Code. 10243391
  • 202.00€ / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
250 mL
Respirometry