Learn More
Hirschmann™ Measuring flask Class A with pre-packing regulation
Achieve precise standard solutions. Hirschmann™ Measuring flask Class A with pre-packing regulation is standardized to contain or to deliver a definite amount of liquid. Flask has blue graduations.
Brand:  Hirschmann™ 2950175
 View more versions of this product
Product Code. 11892582
  • 171.00€ / Pack of 2
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class A
  • Pursuant to the rules of the pre-packing regulation
  • Certified conformity
  • Blue graduation
  • Non-ground, rimmed
  • Printed scale on the neck for simple and rapid identification of deviation of actual value from normal value
  • Calibrated for IN
Specifications
Specifications
50 mL
0.5 mL
Yes
47 to 53 mL
140 mm
A