Learn More
Macherey-Nagel™ Medi-Test™ Glucose
Use test strips for rapid urine screening. Medi-Test™ Glucose test strips provide reliable urinalysis at the point of care.
Brand:  Macherey-Nagel™ 93024
Product Code. 11360415
  • 12.60€ / Pack of 100
In Stock
Description
  • Test strips provide useful information within minutes on the health status of the patient
  • Simplifies the decision about further diagnostic and therapeutic action
  • Also suitable for patient self testing
Specifications
Specifications
100