Learn More
KOLMI Medical Mask, OP-Air
Type II medical mask with earloops
Brand: KOLMI M14311-30
3.43 EUR valid until 2025-06-21
Use promo code "24097" to get your promotional price.
Description
- Medical mask Op-Air
- Earloop EN 14683 Type II
- Made in France
Specifications
Blue | |
EN 14683 | |
Earloops |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
-
Multiple Manufacturers
Outlet Corner
Use code 24097 to receive this promotion.Promo Code: 24097Expires: 2025-06-16
Days
27Hours
4Minutes
34Seconds
55