Learn More
KD Scientific Medium Pressure Nut/Ferrule Auto-fill Kit
Medium Pressure Nut/Ferrule Auto Fill Kit contains Nut PEEK 1/16, 1/4-28 (3 pcs), Ferrule, flangeless 1/16 (3 pcs) and Teflon tubing (5 ft/1.5 m).
Brand: KD Scientific 788347
158.65 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spare part / accessory for KD Scientific Syringe pumps
- Compatible with KD Legacy and Legato™ products
Specifications
None | |
Auto-fill Kit |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Product Content Correction
Product Title
KD Scientific Medium Pressure Nut/Ferrule Auto-fill Kit >