Learn More
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
11719728
|
Fisherbrand™
CALMK22 |
83919 Device |
340.00€
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
11769728
|
Fisherbrand™
11769728 |
FOR 83925 |
351.00€
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||||
DescripciĂłn
- ORP electrode Fisherbrand 1m cable with BNC epoxy body 12mm x 120mm
- ORP Electrode with BNC Epoxy Body
- Combined ion-sensitive electrode with solid membrane interchangeable flat surface
- The ion to be detected passes through the membrane generating a difference of potential proportional to ionic concentration
- Analysis time generally less than one minute
- Sample preparation steps such as filtering or distillation rarely necessary
- Calibration should be carried out as often as possible (less once a day) at the same temperature as the sample
- Measurement temperature 0 to 50 ° C
ÂżDetecta una oportunidad de mejora?Comparta una correcciĂłn de contenido
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂtulo del producto
Fisherbrand™ Ion Selective Membrane Replacement Kits