Learn More
Emma Safety Footwear Mendoza Safety Shoes
State-of-the art Sympatex™ construction provides perfect foot protection for hands-on roles within the Chemical Industry and Agriculture. Virtually waterproof, the Mendoza boots allow feet to breathe properly, keeping them cool and dry.
Brand:  Emma Safety Footwear MM58284848
 View more versions of this product
Product Code. 15918791
  • 227.00€ / 1 pair
Estimated Shipment: 20-05-2025
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck / Sympatex™ construction
  • Waterproof
  • GripForce™ Duo Heat Resistant Outsole
Specifications
Specifications
EN ISO 20345:2011
Mendoza
Oiled Nubuck
Steel
Brown
Hydro-Tec™ Sanitized Silver / Sympatex™
48