Learn More
Emma Safety Footwear Mendoza Safety Shoes
State-of-the art Sympatex™ construction provides perfect foot protection for hands-on roles within the Chemical Industry and Agriculture. Virtually waterproof, the Mendoza boots allow feet to breathe properly, keeping them cool and dry.
Brand: Emma Safety Footwear MM58284842
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck / Sympatex™ construction
- Waterproof
- GripForce™ Duo Heat Resistant Outsole
Specifications
EN ISO 20345:2011 | |
Mendoza | |
Oiled Nubuck | |
Steel |
Brown | |
Hydro-Tec™ Sanitized Silver / Sympatex™ | |
42 |