missing translation for 'onlineSavingsMsg'
Learn More
Chrysene in Acetonitrile 10μg/mL, Fisher Chemical™ Product Code.: 12982534
compliance-icons compliance-icons

Chrysene in Acetonitrile 10μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12982534 Shop All Fisher Chemical Products
10 mL
Click to view available options
Quantity:
10 mL
This item is not returnable. View return policy

Product Code. 12982534

Brand: Fisher Chemical P813030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 218-01-9
Molecular Formula C18H12
Concentration 10 ÎĽg/mL
UN Number 1648
InChI Key WDECIBYCCFPHNR-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43
IUPAC Name chrysene
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 228.294
PubChem CID 9171
Quantity 10 mL
ChEBI CHEBI:51687
Chemical Name or Material Chrysene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Chrysene 10ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chrysene in Acetonitrile 10μg/mL, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.