Learn More
Lauda™ Hose with Multi-layer Thermal and Cold Insulation
Material: Metal
Brand:  Lauda™ LZM 049
Product Code. 10242272
  • 1085.00€ / Each
Estimated Shipment: 09-01-2025
Description
  • Multi-layer
  • For connections with thread M16X1
Specifications
Specifications
-60°C to +350°C
Metal
34 mm
Hose
200 mm
10 mm