Learn More
Eppendorf™ Metering, For Varitip S Dispensing Part
Aspirate liquid from tall, narrow-necked vessels. Eppendorf™ Varitip™ S Forms a system with the Eppendorf Maxitip for aspirating 2.5 to 10mL with Eppendorf Varipette.
Brand: Eppendorf™ 0030050533
92.70 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Made of highest quality, virgin polypropylene (PP) free of biocides, plasticizers and latex
- Available in packs of 30
- Air displacement principle
- No carryover
- Ensures drip-free dispensing of liquids with a high vapor pressure
Specifications
Eppendorf Maxitip and Varipette | |
30 Pack |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92% -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31