missing translation for 'onlineSavingsMsg'
Learn More
Methyl 2-furoate, 98+% Product Code.: 11457876

Methyl 2-furoate, 98+%

Product Code. 11457876
100 g, 100g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11457876

Brand: Thermo Scientific Alfa Aesar A14009.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-13-2 | C6H6O3 | 126.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-13-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00003236
InChI Key HDJLSECJEQSPKW-UHFFFAOYSA-N
Synonym methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl esterShow MoreShow Less
PubChem CID 11902
IUPAC Name methyl furan-2-carboxylate
SMILES COC(=O)C1=CC=CO1

Specifications

Density 1.179
Boiling Point 180°C to 181°C
Flash Point 73°C (163°F)
Refractive Index 1.487
Quantity 100 g
UN Number UN2810
Beilstein 111110
Merck Index 14,4307
Formula Weight 126.11
Percent Purity ≥98%
Chemical Name or Material Methyl 2-furoate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245107 Certificate TypeCertificate of Analysis Date23/12/2022 Catalog NumberA14009.14,A14009.36,A14009.22
Lot Number10242280 Certificate TypeCertificate of Analysis Date16/08/2022 Catalog NumberA14009.14,A14009.36,A14009.22
Lot Number10232187 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14009.14,A14009.36,A14009.22
Lot Number10150650 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14009.14,A14009.36,A14009.22
Lot Number10230341 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14009.14,A14009.36,A14009.22
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 2-furoate, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.