Learn More
Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals Product Code.: 11401497
Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals
Product Code. 11401497
500 g
Click to view available options
Quantity:
100 g
500 g
Product Code. 11401497
Brand: Thermo Scientific Alfa Aesar B24953.36
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14205-39-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00008072,MFCD00008072,MFCD00008072
InChI Key XKORCTIIRYKLLG-ONEGZZNKSA-N
Synonym methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl esterShow MoreShow Less
PubChem CID 643918
IUPAC Name methyl (Z)-3-aminobut-2-enoate
SMILES COC(=O)\C=C(/C)N
Specifications
Melting Point 81°C to 85°C
Boiling Point 122°C (42 mmHg)
Flash Point 91°C (195°F)
Quantity 500 g
Beilstein 956592
Sensitivity Hygroscopic
Formula Weight 115.13
Percent Purity 97%
Chemical Name or Material Methyl 3-aminocrotonate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0386842 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberB24953.22,B24953.36
Lot NumberA0343963 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberB24953.22,B24953.36
Lot Number10177347 Certificate TypeCertificate of Analysis Date04/07/2023 Catalog NumberB24953.22,B24953.36,B24953
Lot Number10150271 Certificate TypeCertificate of Analysis Date23/06/2023 Catalog NumberB24953.22,B24953.36,B24953
Lot Number10170766 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24953.22,B24953.36
Product Title
Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals >