Learn More
Methyl 5-methylnicotinate, 94%, Thermo Scientific Chemicals
Description
5-Methyl-N?-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N?-nitrosonornicotine is used as the starting material in organic compound synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5-Methyl-N′-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N′-nitrosonornicotine is used as the starting material in organic compound synthesis.
Solubility
Soluble in Chloroform and Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 42°C to 46°C |
Boiling Point | 90°C (3 mmHg) |
Quantity | 1 g |
Beilstein | 472498 |
Synonym | methyl 5-methylnicotinate, methyl5-methylnicotinate, 5-methylnicotinic acid methyl ester, 5-methyl-nicotinic acid methyl ester, methyl 5-methyl nicotinate, 3-pyridinecarboxylic acid, 5-methyl-, methyl ester, 3-methoxycarbonyl-5-methylpyridine, 5-methylpyridine-3-carboxylic acid methyl ester, 5-methylnicotin methyl ester, acmc-1clej |
Solubility Information | Soluble in Chloroform and Methanol. |
Formula Weight | 151.17 |
Percent Purity | 94% |
Chemical Name or Material | Methyl 5-methylnicotinate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.