missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange 1 g/L in Water Indicator Solution, Chem Lab

Brand:  Chem Lab CL06.1302.0250

Product Code. 11993333

  • 26.20€ / 250mL
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Orange
100°C
1 g/L
327.34
Standard test method: ASTM D-974, D-664, D-3712
250 mL
547-58-0
0°C
1 g/mL
Polyethylene
3.1 to 4.4
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl Orange 1 g/L in Water Indicator Solution, Chem Lab > Methyl Orange 1 g/L in Water Indicator Solution, 250 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.