missing translation for 'onlineSavingsMsg'
Learn More
Methylurea, 98%, Thermo Scientific Chemicals Product Code.: 11335045

Methylurea, 98%, Thermo Scientific Chemicals

Product Code. 11335045
1000 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11335045

Brand: Thermo Scientific Alfa Aesar B24772.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-50-5
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.083
MDL Number MFCD00007950
InChI Key XGEGHDBEHXKFPX-UHFFFAOYSA-N
Synonym 1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
PubChem CID 11719
ChEBI CHEBI:44383
IUPAC Name methylurea
SMILES CNC(=O)N

Specifications

Melting Point 96°C to 101°C
pH 6.7
Odor Odorless
Quantity 1000 g
Beilstein 878189
Formula Weight 74.08
Percent Purity 98%
Chemical Name or Material Methylurea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methylurea, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.