Learn More
Metro™ Starsys™ Cart Accessories
Parts for use with Starsys carts
Brand: Metro™ B5PC-TL
This item is not returnable.
View return policy
Description
- 5 in. (127mm)
- Total lock
- All polymer
- Recommended for corrosive environments
Specifications
Stem Caster | |
Metro Starsys cart | |
Polymer |
For corrosive environments | |
Total lock |