Learn More
Metro™ Starsys™ Overhead Storage Unit
Microban™ antimicrobial protection prevents growth of stains and odor-causing bacteria.
1495.00€ - 3450.00€
Specifications
Color Taupe
Depth (English) Exterior 13.8 in.
Height (English) Exterior 29.5 in.
Product Type Overhead Storage Unit
Height (English) 29.5 in.
View More Specs
Products 7
Product Code Brand Width (English) Door Style Price Quantity & Availability  
Product Code Brand Width (English) Door Style Price Quantity & Availability  
15917271
View Documents
Metro™
SXRSOH27P2N
21.5 in. No Door
1495.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
15907271
View Documents
Metro™
SXRSOH27P2C
21.5 in. Fixed Window
2140.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
15987241
View Documents
Metro™
SXRDOH27P2C
41.3 in. Fixed Window
3360.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
15927271
View Documents
Metro™
SXRSOH27P2S
21.5 in. Single
1835.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
15997241
View Documents
Metro™
SXRDOH27P2N
41.3 in. No Door
1990.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
15917251
View Documents
Metro™
SXRDOH27W2C
41.3 in. Double, Fixed Window
3450.00€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
15907251
View Documents
Metro™
SXRDOH27P2S
41.3 in. Double
2785.00€
Each
Estimated Shipment: 23-05-2025
Description
  • Built on same widths as Starsys base units utilizing very similar construction
  • Insures that wall cabinets align with lower product configuration in application
  • All overhead cabinets include corrugated polymer inner panels in their assembly as well as clean-design back panel, molded with cosmetic side facing inside of cabinet, creating much cleaner appearance - especially when used without doors or in conjunction with clear door
  • Necessary wall mount rails/hardware are included with each wall cabinet
  • Microban™ antimicrobial protection inhibits growth of stain and odor-causing bacteria on Starsys, keeping surface areas cleaner between cleanings
Specifications
Specifications
Taupe
29.5 in.
29.5 in.
13.8 in.
Overhead Storage Unit