Learn More
Mettler Toledo™ uPlace™ Electrode Holder for SevenCompact™ Meters
For use with SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter.
Brand: Mettler Toledo™ 30019823
240.54 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- uPlace electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample
- This allows for faster measurements and reduces the risk of the sample vessel tipping over or damaging the sensor
Specifications
uPlace Electrode Holder | |
uPlace electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample | |
SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%