Learn More
Mettler Toledo™ Replacement Cap for InLab™ OptiOx™ Optical DO Sensor
For InLab OptiOx Optical DO Sensor.
Brand:  Mettler Toledo™ 51344630
242.25 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 11917811
  • 255.00€ / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
In Stock
Description
For InLab OptiOx Optical DO Sensor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OptiOx replacement Cap
Replacement Cap
OptiOx replacement Cap
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
Product Content Correction
Product Title
Mettler Toledo™ Replacement Cap for InLab™ OptiOx™ Optical DO Sensor > Replacement cap