missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies mFreSR™

Defined, serum-free cryopreservation medium for human ES and iPS cells

Encompass_Preferred

525.15€ - 633.15€

Specifications

Cell Type Pluripotent Stem Cells
For Use With (Application) Cryopreservation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13479008
Encompass_Preferred
View Documents
STEMCELL Technologies
05855
50 mL
525.15€
50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15522640
Encompass_Preferred
View Documents
STEMCELL Technologies
05854
10 x 5 mL Tubes
633.15€
50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • mFreSR™ is a serum-free cryopreservation medium designed for the cryopreservation of human embryonic and induced pluripotent stem cells (ES and iPS cells)
  • mFreSR™ contains DMSO and is complete and ready-to-use
  • Together with mTeSR™1, TeSR™2, or mTeSR™ Plus, mFreSR™ eliminates the use of feeders and serum
  • Human ES and iPS cells cryopreserved in mFreSR™ have thawing efficiencies higher than reported conventional thawing methods using serum
Specifications

Specifications

Pluripotent Stem Cells
Cryopreservation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies mFreSR™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.