Learn More
uvex Micro Black/Orange PU/PU 65092 Boot - Width 11
Optimal shock absorption, ergonomic fit, low weight and excellent climate characteristics ensure max performance at work and redefine the limits for safety shoes; range in protection classes S3 and S1 P
149.00€
Specifications
Color | Black, Orange |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17220931
|
uvex
6509235 |
35 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17230931
|
uvex
6509236 |
36 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17240931
|
uvex
6509237 |
37 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17250931
|
uvex
6509238 |
38 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17260931
|
uvex
6509239 |
39 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17270931
|
uvex
6509240 |
40 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17280931
|
uvex
6509241 |
41 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17290931
|
uvex
6509242 |
42 |
149.00€
1 pair |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
17200941
|
uvex
6509243 |
43 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17210941
|
uvex
6509244 |
44 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17220941
|
uvex
6509245 |
45 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17230941
|
uvex
6509246 |
46 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17240941
|
uvex
6509247 |
47 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17250941
|
uvex
6509248 |
48 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17260941
|
uvex
6509249 |
49 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17270941
|
uvex
6509250 |
50 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17280941
|
uvex
6509251 |
51 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
17290941
|
uvex
6509252 |
52 |
149.00€
1 pair |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
Description
Uvex 2 Boot 6509.2 S3 SRC
- Sporty, multifunctional safety boot
- Extremely light and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity and anti-magnetic properties
- Metal-free, penetration-resistance insole in accordance with latest standard, which does not impact flexibility
- Made with new high-performance technology for exceptional wearer comfort
- Uvex Climazone technology for optimum comfort
- Breathable, water-repellent high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic textiles, suitable for those allergic to chrome
- Reflecting elements for more safety
- Virtually seam-free construction to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue, dust tongue
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Uvex topshield scuffcap to protect the upper and extend its life
- Fulfills ESD requirements: resistance to ground Kleiner of 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6509.2
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane (PU/PU) | |
Reusable | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S3 | |
ESD Safety Shoe |
Product Title
uvex Micro Black/Orange PU/PU 65092 Boot - Width 11