Learn More
Socorex™ Acura™ Self-refill 865 Microdispensing Pipette
Hand-held microdispenser intended for repeat distribution of microliter volumes. Socorex™ Acura™ Self-refill 865 Microdispensing Pipette includes a three-way valve system for automatic self-refilling.
Brand:  Socorex™ 865.0200
Product Code. 10509022
  • 457.00€ / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hand-held.
  • Three-way valve system for automatic self-refilling.
  • Durable and resistant to aggressive media.
  • Excellent ergonomics.
  • Ultra-soft plunger activation.
  • Large volume display.
  • Versatile feeding through bottle, tubing or syringe.
  • Easy in-lab calibration.
  • Individually serial numbered and Q.C. certified.
  • Autoclavable.
Specifications
Specifications
<±2.5% - <±1.0%
1
Microdispenser
<1.5% - <0.3%
36 Months
Autoclavable
Acura
20 to 200 μL
Aggressive media
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Socorex™ Acura™ Self-refill 865 Microdispensing Pipette > Volume: 20-200μL