Learn More
Ansell MICROFLEX™ LifeStar™ EC 93-868 High Risk Nitrile Exam Gloves
Product Code. 16875440
Click to view available options
Size:
7
8
9
10
11
12
Dual-layered gloves provide two levels of reliable barrier protection against hazardous substances and high-risk patient handling.
  • Two layers of reliable barrier protection
  • For protection against hazardous substances and high-risk patient handling
  • Designed with non-stick and non-foaming properties
  • Reduced interference and increased productivity
  • Advanced barrier protection for demanding tasks
  • AQL 0.65
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 455 1-4, EN 420:2003 + A1:2009
Color White
Disposable Single-use
Material Nitrile
Product Type Nitrile Examination Glove
Size 10
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKPT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
For Use With (Application) Handling hazardous substance and high-risk patients
Glove Cuff Style Beaded
Series 93-868