Learn More
Thermo Scientific™ Sealing Tape for 96-Well Plates
Clear, self-sticking sealing tape for standard microplates.
Brand:  Thermo Scientific™ 15036
118.58 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
Product Code. 10105383
  • 133.00€ / Pack of 100
In Stock
Description
Clear, self-sticking sealing tape for standard microplates.
Thermo Scientific Microplate Sealing Tape is a package of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.
Features of Microplate Sealing Tape:
• Pressure-sensitive—position the tape over plate and press to seal; easy to remove
• Separate sheets—each sheet of tape is supplied on its own easy-to-peal backing (liner)
• Durable and inert—film thickness provides support and has a thin layer of acrylic adhesive
Use the disposable sheets to seal and reseal a microplate two or three times for incubation steps in an ELISA or protein assay procedure. Seal plates quickly and easily to allow mixing or prevent evaporation during assay incubation steps.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Non-sterile
100 sheets
Pressure Sensitive
Polyester
96/384
Film, Seal, or Applicator
96/384-well Plate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberVC298114 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number15036
Lot NumberZK402058 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number15036
Lot NumberZK402056 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number15036
Lot NumberZK402057 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number15036
Lot NumberZJ401875 Certificate TypeCertificate of Analysis Date02/11/2024 Catalog Number15036
Special Offers
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Sealing Tape for 96-Well Plates >