Learn More
Karl Hecht™ Assistent™ Microscope Slides with Single Cavity
Holds specimens such as liquids and cultures for observation with a microscope. Karl Hecht™ Assistent™ Microscope Slides with Single Cavity are manufactured from clear glass with ground edges.
Brand: Karl Hecht™ 42410010
This item is not returnable.
View return policy
Description
- Approx. 1.5mm thick
- Dimensions: approx. 76 x 26mm
- Supplied in packs of 50
Specifications
Microscope Slide | |
Clear | |
1.5 mm | |
76 x 26 mm |
1 | |
Ground Glass | |
50 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Karl Hecht™ Assistent™ Microscope Slides with Single Cavity > Product Type: Microscope Slide