Learn More
Carl Zeiss™ Microscope Stage Micrometers
Finely divided stage micrometer for calibrating optical systems with eyepiece graticule patterns. Carl Zeiss™ Microscope Stage Micrometer includes 1mm and 1/100mm intervals.
Brand:  Carl Zeiss™ 474027-0000-000
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11866422
  • 468.52€ / Each
Estimated Shipment: 13-11-2024
to see stock.
Add to basket
Includes: 5 lines with 1mm intervals in y-direction, 100 lines with 1/100mm intervals in y-direction
Description
Description
  • For calibrating optical systems with eyepiece graticule patterns
  • 5 lines with 1mm intervals in y-direction
  • 100 lines with 1/100mm intervals in y-direction
Specifications
Specifications
Stage Micrometer
Reflected light microscope, eyepiece graticule
Stage micrometer, 5 lines with 1mm intervals in y-direction