Learn More
Carl Zeiss™ Microscopic Stand
Use these stands with Zeiss™ microscopes.
Brand:  Carl Zeiss™ 430037-9010-000
Product Code. 11865053
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Microscope stand Axio Lab.A1 HAL 35, 5x H
  • Z-drive with fine drive knob left and fine drive disk right, flat with scala - nosepiece 5x bright field, M27
  • Integrated 12V DC 50W power unit, stabilized 100 to 240V AC/50 to 60Hz/110VA
  • Transmitted-light illumination with halogen reflector lamp 12V 35W, LED optional
  • Stage and condenser carrier - country-specific power cable
Specifications
Specifications
Microscopic Stand
1 Ea.