missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer Mighty Small precast gels
Hoefer Mighty Small precast gels are high-performance mini polyacrylamide gels commonly used for protein separation.
Brand: Hoefer MSG010F
This item is not returnable.
View return policy
Description
- Specification:
- Plate Size: 10 x 8 cm
- Gel Size: 8.5 x 7 cm, 1 mm thickness
- Wells: 15 well (12 wells available)
- System/Buffer: Bis-Tris System, MOPS Buffer
- Loading Volume: For 15 wells: max 30 μl (recommended: ≤ 15 μl)
- Package: 10 pcs/box
- Weight:0.66 lb.
- Storage temp: 2 to 8°
- Shelf life: 12 months
- Advantages:
- upgraded Bis-tris system with moderate PH to ensure less modification on the protein to get a sharp and clear band
- Save trouble and time from making gels
Specifications
MOPS | |
10% | |
10% | |
1.0 mm | |
7 | |
25 ÎĽL | |
10 to 300 kDa | |
15-well |
Clear | |
Mini Gel Tank | |
Mini | |
Bis-Tris | |
Precast gels | |
Protein | |
Denaturing | |
10 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hoefer Mighty Small precast gels > 10% Bis-Tris, 15 wells
Spot an opportunity for improvement?Share a Content Correction