Learn More
Merck Millicell ÎĽ-Angiogenesis Inhibition Assay Kit
Brand: Merck MMA125
This item is not returnable.
View return policy
Description
Order Info
dimType
Specifications
Fluorescence | |
For Real-Time Monitoring of Changes In Tubule Formation With Unprecedented Optical Resolution | |
Millicell™ μ-Angiogenesis Slides: Five tissue Culture Treated Slides, Each Slide Contains 15 wells, D, L-Sulforaphane: One 1 mg vial, Calcein AM: One 50 μg vial, One 1 mL vial | |
1 Kit |
Product Content Correction
Product Title
Merck Millicell ÎĽ-Angiogenesis Inhibition Assay Kit > Millicell Micrometer-Angiogenesis Inhibition Assay Kit