Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™
Versatile check valves can be installed in any position
Brand:  Cole-Parmer 50115-22
 View more versions of this product
Product Code. 30054160
  • 4.80€ / Each
Estimated Shipment: 06-11-2025
Description
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressure
Specifications
Specifications
0.3125 in.
5/16 in. hose barb
Viton
0.13
Barbed
Product Title
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™ > 5/16 in. hose barb