Learn More
Greiner Bio-One MiniCollect™ Tube, Gold Cap, CAT Serum Sep Clot Activator, Non-Sterile
Includes inner wall coated with microscopic silica particles to activate coagulation and a separator gel to separate serum and blood cells.
Brand:  Greiner Bio-One 450533
Product Code. 15778169
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
  • Serum separator tubes contain a separator gel in the base of the tube; during centrifugation, this gel forms a stable barrier between the serum and the blood cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CAT Serum Sep Clot Activator/Gel Separator
0.5/0.8 mL
Gold
Non-sterile
Yes
500/Pk.