Learn More
Greiner Bio-One MiniCollect™ Tube, Light-Blue Cap, 9NC Coagulation Sodium Citrate 3.2%, Sterile
Contains a buffered sodium citrate solution to aid in examination of coagulation parameters.
Brand: Greiner Bio-One 450538
This item is not returnable.
View return policy
Description
Contains a buffered sodium citrate solution to aid in examination of coagulation parameters.
- This light blue top sodium citrate tube, or coagulation tube, is used for examination of coagulation parameters.
- Coagulation tubes contain a buffered sodium citrate solution.
Specifications
9NC Sodium Citrate 3.2% | |
0.5 mL | |
Light Blue |
Sterile | |
Yes | |
500/Pk. |