missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MIP-1 beta Human ProQuantum Immunoassay Kit
GREENER_CHOICE

Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument

3350.00€

Specifications

Accession Number P13236
Assay Range 0.064 to 5000 pg/mL
Assay Sensitivity <0.05 pg/mL
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17183109
GREENER_CHOICE
View Documents
Invitrogen™
A355975
5 x 96 Tests
3350.00€
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

  • High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
  • Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
  • Fast, easy workflow: No wash steps, 2 hours from sample to answer
  • Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
  • No proprietary instrument to purchase: Runs on any real-time PCR instrument
  • ProQuantum software available: Robust data analysis and statistical group wise comparison
Specifications

Specifications

P13236
<0.05 pg/mL
Proximity-based Immunoassay
Human
qPCR Instruments
ACT2; AT744.1; G-26; HC21; LAG-1; LAG1; MIP-1-beta; MIP1B; MIP1B1; SCYA2; SCYA4
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard
RUO
Frozen at -20°C
30 min.
0.064 to 5000 pg/mL
Taqman™ Real-time PCR
ProQuantum
Protein quantitation
6351
1 hr.
FAM™
Serum,2-5μL; Supernatant,2-5μL; Plasma,2-5μL
Human
2 hr.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.