Learn More
Cytiva Mixed Cellulose Ester Filtration Membrane
These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters
Brand: Cytiva 10401664
This item is not returnable.
View return policy
Description
Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters. Also, the color contrast provided by the filter surface facilitates particle detection and minimises eye fatigue. The ME range has a lower cellulose acetate content compared to the WME range of membranes. Circles, gridded.- Sterile options available for critical applications
- Excellent contrast for easier particle detection
- Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
- Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
- The membrane offers a high degree of internal surface area for greater adsorption of product
- Higher dirt loading capacity
- Biologically inert with good thermal stability
- No surfactants to contaminate samples
- Uniform microporous structure of membrane gives high flow rates
Specifications
Autoclavable | |
0.45 μm | |
12.5/100/12.5sec./mL/cm2 | |
Plain | |
100Pack |
MCE | |
Non-sterile | |
Microbiological Colony Counting Procedures | |
ME25 | |
Membrane Filter |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ||Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose