Learn More
Tocris™ ML 335
Selective K2P2.1 (TREK-1) and K2P10.1 (TREK-2) activator
Brand:  Tocris™ 6887/50
482.93 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18705796
  • 623.00€ / 50mg
Estimated Shipment: 28-11-2024
Description
ML 335 is a selective K2P2.1 (TREK-1; KCNK2) and K2P10.1 (TREK-2; KCNK10) activator (EC50 values of 14.3 μM and 5.2 μM, respectively); directly stimulates the C-type gate via the K2P modulator pocket. Does not activate K2P4.1 (TRAAK).
Specifications
Specifications
50 mg
373.25
98%
C15H14Cl2N2O3S
Activator