Learn More
Motic™ MLC-150 Cold Light
Halogen light source with corded remote control for ease of use in all lab settings.
Brand:  Motic™ 1101002000011
Product Code. 11776576
  • 462.00€ / Each
Estimated Shipment: 05-05-2025
Description
  • 21V/150W (EKE) Halogen illumination
  • Local/remote switch for intensity control
  • 2m remote control intensity
  • Color temperature LED read out
Specifications
Specifications
Illuminator
Remote switch
150 w
Halogen
21 V
Certificates
View accessory products
Product Title
Motic™ MLC-150 Cold Light > MLC-150 cold light