Learn More
Elma™ Modular Baskets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
13.85€ - 41.00€
Specifications
Type | Small Basket |
---|---|
For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
Product Code | Brand | Diameter (Metric) | Height (Metric) | Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Style | Price | Quantity & Availability | ||||
17851337
|
Elma™
3000006017 |
21 mm | 12 mm | 4 Sections |
13.85€
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
17841337
|
Elma™
3000006018 |
64 mm | 2 mm | 9 Sections |
13.85€
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
17821337
|
Elma™
3000006012 |
64 mm | 16 mm | 1 Section |
21.55€
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
17821347
|
Elma™
3000006013 |
64 mm | 16 mm | 5 Sections |
35.90€
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
17811347
|
Elma™
3000009104 |
64 mm | 42 mm | 1 Section |
41.00€
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
Description
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Specifications
Small Basket | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
Product Title
Elma™ Modular Baskets