missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ TV100YK-MODSYS TV Complete Vertical Modular System
Provide a fully integrated solution within one gel tank for both twodimensional (2-D) electrophoresis and electroblotting.
Brand: Fisherbrand™ TV100YK-MODSYS
This item is not returnable.
View return policy
Description
- Comprises of one standard GRM, one CAPGRM and one EBGRM, which are interchangeable within the same gel tank, allowing the user to perform PAGE, IEF and electroblotting either independently or as part of a fully integrated system
- CAPGRM (capillary gel running module) for first dimension electrophoresis of up to 10 ampholyte capillary gels, a maximum 8 or 17cm in length and 1 or 4mm in internal diameter
- Twin-plate standard GRM is designed specifically for SDS-PAGE techniques requiring self-cast gels
- Twin-cassette EBGRM (Electroblotting Gel Running Module) allows simultaneous transfer of 2 polyacrylamide gels in colour-coded electroblotting cassettes
- Plate electrodes which are only 6cm apart maximise the intensity of the electric field
- Casting base and cooling are included as standard - act in tandem with glass plate stops within the GRM to lock the plates firmly in position on the casting base
- Snap-lock connectors allows the cooling coil within the tank to be connected to an external chiller unit for high voltage/current separations
Specifications
Cooling coil connected to an external chiller unit | |
Cooling coil within the tank to be connected to an external chiller unit | |
Twin-plate mini-gel unit with GRM and cooled gel tank, with built-in cooling coil and quick-fit tubing, lid, 2 x (10W x 10cmH) plain glass plates, 2 x (10 x 10cm) notched glass plates,1 x dummy plate, 4 x 1mm thick spacers, 2 x spacer aligners and 2 x 1mm thick 12-sample combs, casting base and 2 x silicone seals; CAPGRM, 10 x 1mm glass capillaries, 4 x 1.5mm thick spacers, 2 x 1.5mm thick 1-D preparatory combs, 10 x blanking ports and 1 x gel extraction platform; plus EBGRM, 2 x electroblotting cassettes and 4 x fibre pads | |
0.1 cm | |
Inner Buffer Chamber: 90 mL | |
15 cm |
1 to 1.75 hr. | |
W x H: 8 x 8.5 cm | |
Plate: 0.2 cm | |
Gel Tank: 1600 mL | |
W x H: 28 x 18 cm | |
100 to 150 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction