missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele Module A306/1 4 éprouvettes 1 ou 2L
Brand: Miele 12683100
1830.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Module A306/1 4 éprouvettes 1 ou 2L
This item is not returnable.
View return policy
Description

Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Miele Module A306/1 4 éprouvettes 1 ou 2L >
Spot an opportunity for improvement?Share a Content Correction