Learn More
Molecular Sieve Dehydrate Fluka with indicator for drying solvents, Honeywell Fluka™
BEADS, WITH INDICATOR FOR DRYING SOLVENTS
146.06€ - 527.34€
Specifications
Chemical Name or Material | Molecular Sieve Dehydrate Fluka with indicator for drying solvents |
---|---|
CAS | 7646-79-9, 1318-02-1 |
MDL Number | MFCD00147630 |
UN Number | UN3077 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15646740
|
Honeywell-Fluka
69839-250G |
250 g |
146.06€
250g |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
15636740
|
Honeywell-Fluka
69839-1KG |
1 kg |
527.34€
1kg |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Molecular Sieve Dehydrate Fluka with indicator for drying solvents | |
7646-79-9, 1318-02-1 | |
MFCD00147630 | |
UN3077 |
Safety and Handling
P201-P261-P280-P284-P305 + P351 + P338 + P310-P308 + P313