missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl fumarate, Thermo Scientific™

Brand:  Thermo Scientific Acros 469750050

 View more versions of this product

Product Code. 17760669

  • 48.30€ / 5g
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Monomethyl fumarate
5 g
No special requirements
C5 H6 O4
White to Almost-white
2756-87-8
145°C to 150°C
Building blocks
Powder or Crystals
≥96.0%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers