Learn More
Motic™ Moticam A1
The Moticam A1 is ideal for newcomers and advanced enthusiasts who are looking for an affordable digital microscope instrument. Capture everything you need at 720p and work at a steady frame rate with all accessories included.
Brand: Motic™ 1100600101412
This item is not returnable.
View return policy
Description
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
- Live Display Modes: 1280 x 720 px, 1024 x 768 px
Specifications
Camera | |
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap | |
1/3.1 in. |
Moticam A | |
sCMOS | |
30 fps |
Product Content Correction
Product Title
Motic™ Moticam A1 > Moticam A1; Frequency (Frame Rate): 30 fps; Sensor Types: sCMOS; Product Line: Moticam A Series; Includes: Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap