missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Armenian and Syrian Hamster IgG1, Clone: HIG-632, Secondary Antibody, BD

Brand:  BD Biosciences 550637

398.46 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15856358

  • 399.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Based on ELISA, the HIG-632 antibody reacts specifically with Armenian and Syrian hamster IgG1 monoclonal antibodies. The HIG-632 mAb does not react with other hamster IgG groups or hamster IgM.

Host Species: Mouse
Clone: HIG-632
Isotype: IgG2b Îş
Species Reactivity: Hamster
Immunogen: Pooled Armenian Hamster IgG mAb

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse Anti-Armenian and Syrian Hamster IgG1
HIG-632
Mouse
0.5 mg
Immunology
Hamster
Monoclonal
0.5mg/mL
Affinity Purified
RUO
Secondary
IgG2b Îş
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mouse Anti-Armenian and Syrian Hamster IgG1, Clone: HIG-632, Secondary Antibody, BD > 0.5mg; Unlabeled

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.